Skip to Content
Merck
All Photos(1)

Key Documents

04721

Supelco

Abberior® STAR 488, streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

manufacturer/tradename

Abberior GmbH

fluorescence

λex 503 nm; λem 524 nm±10 nm in H2O

suitability

corresponds for SDS-PAGE

detection method

fluorometric

storage temp.

2-8°C

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

Unconjugated dye ≤5% of total fluorescence

Legal Information

abberior is a registered trademark of Abberior GmbH

related product

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service