Skip to Content
Merck
All Photos(1)

Key Documents

41270

Millipore

Violet Red Bile Lactose Dextrose Agar

suitable for microbiology

Synonym(s):

VRB Lactose Dextrose Agar, Violet Red Bile Lactose Glucose Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
bile salts, 1.5 g/L
crystal violet, 0.002 g/L
dextrose monohydrate, 10 g/L
lactose monohydrate, 10.0 g/L
neutral red, 0.03 g/L
pancreatic digest of gelatin, 7.0 g/L
sodium chloride, 5.0 g/L
yeast extract, 3.0 g/L

pH

7.4±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

selective and differential for Citrobacter spp.
selective and differential for Enterococcus spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

General description

A selective Agar for the detection of Enterobacteriaceae according the European Pharmacopeia. he medium is specially used in the recovery of process-stressed bacteria using a progressive enrichment technique.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service