Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BCR067

Quartz (particle size)

BCR®, certified reference material, 2.40-32.00 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

BCR®

form

particles

manufacturer/tradename

JRC

particle size

2.40-32.00 μm

application(s)

glass & ceramic

format

neat

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR067

Legal Information

BCR is a registered trademark of European Commission

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service