PHR8798
Lacosamide Related Compound F

certified reference material, pharmaceutical secondary standard
Synonym(s):
2-Acetamido-N-benzyl-3-hydroxypropionamide, lacosamide ep impurity f
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
agency
traceable to EP Y0001989
traceable to USP 1357170
API family
lacosamide, lacosamide
packaging
pkg of 50MG
storage temp.
-10 to -25°C
Looking for similar products? Visit Product Comparison Guide
Other Notes
Footnote
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service