Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1983

Sigma-Aldrich

AURORA A, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AIK, ARK1, AURA, AURKA, BTAK, STK15, STK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

62-84 nmol/min·mg

mol wt

~72 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AURKA(6790)

Biochem/physiol Actions

AURORA A belongs to a multigenic family of mitotic serine/threonine kinases which are involved in the control of chromosome segregation. AURORA A is involved in centrosome separation, duplication and maturation as well as in bipolar spindle assembly and stability. AURORA A is expressed and active at the highest level during G2-M phase of the cell cycle. Overexpression of AURORA A has been found to be correlated with the grade of various human solid tumours. Ectopic AURORA A overexpression in any culture cell line leads to polyploidy and centrosome amplification.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Dutertre et al.
Oncogene, 21(40), 6175-6183 (2002-09-06)
Mammalian aurora-A belongs to a multigenic family of mitotic serine/threonine kinases comprising two other members: aurora-B and aurora-C. In this review we will focus on aurora-A that starts to localize to centrosomes only in S phase as soon as centrioles
Hiroshi Katayama et al.
Cancer metastasis reviews, 22(4), 451-464 (2003-07-30)
Aurora kinases representing a novel family of serine/threonine kinases have been identified as key regulators of the mitotic cell division process. The three members of this kinase family, identified so far, referred to as Aurora-A, Aurora-B and Aurora-C kinases, are
Anna A Kiseleva et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(13), 4179-4193 (2019-03-15)
For many tumors, signaling exchanges between cancer cells and other cells in their microenvironment influence overall tumor signaling. Some of these exchanges depend on expression of the primary cilium on nontransformed cell populations, as extracellular ligands including Sonic Hedgehog (SHH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service