Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV32383

Sigma-Aldrich

Anti-SIX6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Sine oculis homeobox homolog 6 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

guinea pig, bovine, mouse, human, dog, rat, sheep, horse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SIX6(4990)

General description

SIX6 is a homeobox protein and may regulate eye development. Studies have linked homozygous truncations in SIX6 to complex microphthalmia in humans.
Rabbit Anti-SIX6 antibody recognizes chicken, bovine, canine, zebrafish, human, mouse, rat, and pig SIX6.

Immunogen

Synthetic peptide directed towards the N terminal region of human SIX6

Application

Rabbit Anti-SIX6 antibody can be used for western blot (0.5μg/ml) and IHC (4-8μg/ml) applications.

Biochem/physiol Actions

SIX6 is a member of SIX family. It is the homologue of the chick Six6(Optx2) gene. SIX6 is closely related to SIX3 and is expressed in the developing and adult human retina.

Sequence

Synthetic peptide located within the following region: PAACEALNKNESVLRARAIVAFHGGNYRELYHILENHKFTKESHAKLQAL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homozygous truncation of SIX6 causes complex microphthalmia in humans.
M A Aldahmesh et al.
Clinical genetics, 84(2), 198-199 (2012-11-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service