Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.51
UNSPSC Code:
41105324
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CTCAAGGAGGCAGAAAGGTGAAAGCTGGAGATACTGTGTCATATGTCATCTGTCAGGATGGATCAAACCTCACTGCAAGTCAGAGGGCCTATGCGCCTGAGCAGCTGCAGAAACAGGATAATCTAACCATTGACACCCAGTACTACCTGGCCCAGCAGATCCACCCAGTCGTGGCTCGGATCTGTGAACCAATAGACGGAATTGATGCTGTCCTCATTGCAACGTGGTTGGGACTTGACCCCACCCAATTTAGAGTTCATCATTATCATAAAGATGAAGAGAATGATGCTCTACTTGGTGGCCCAGCACAGCTCACTGATGAAGAGAAATACAGGGACTGTGAAAGATTCAAATGTCCATGCCCTACATGTGGAACTGAGAATATTTATGATAATGTCTTTGATGGTTCGGG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... POLA1(5422)
General description
MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| EHU034181-20UG | 04061831341027 |
| EHU034181-50UG | 04061828309801 |