Key Documents
EP1004000001
Eppendorf® VisioNize Pipette Manager
new
includes VisioNize pipette manager software, AC/DC input 100-240 V (AU;CN;EU;GB;US)
About This Item
UNSPSC Code:
41121800
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 1004000001
General description
VisioNize® pipette manager: VisioNize® pipette manager (touch control panel) with VisioNize® pipette manager software
Features and Benefits
  • Easily enter pipetting volumes and other parameters using the touch panel. Settings are immediately transferred to connected electronic pipettes, allowing you to start working right away
  • Get guidance on pipetting techniques by the software-embedded Liquid Manager when working with viscous, volatile and other challenging liquid types
  • Choose from predefined Eppendorf liquid types (e.g. ethanol or enzymes/antibodies) or create your own
  • Automatically document pipetting activities via Pipetting Records and easily export your recordings as PDF file via USB
  • Manage your pipette fleet more effectively and enhance collaboration with integrated features like Pipette Set Creation or Liquid Type Sharing
  • Easily upgrade Eppendorf Xplorer and Xplorer plus electronic pipettes with the Eppendorf Xplorer connect WiFi module. The WiFi modules comes with shorter battery for an optimal fit.
  • The VisioNize pipette manager is based on the VisioNize touch interface concept that has been extensively tested by customers to reduce learning and adjustment time.
  • This is only the beginning - more features are coming.
  • The graphical user interface is based on the VisioNize® touch interface concept that has been extensively tested by customers to reduce learning and adjustment time
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
VisioNize is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)