Key Documents
EP6002000111
Eppendorf® RackScan b for 2D Coded Tubes
new
AC/DC input 240 V, UK plug
About This Item
UNSPSC Code:
41121800
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 6002000111
General description
The more samples you have, the less you want to scan your vials for cryostorage manually. The Eppendorf RackScan can read complete storage racks with up to 96 different barcoded vials from the bottom. RackScan b UK,scanner for 2D coded tubes in SBS racks and cryostorage boxes, incl. software RackScan Analyzer
Application
Sample tracking
Features and Benefits
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Support material of vials available via Eppendorf Homepage for documentation purposes
  • Plug & Transfer with Eppendorf eLABInventory and eLABJournal software for comfortable data transfer
  • RackScan s can be attached as accessory to read barcodes on side labels
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)