Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA007200

Sigma-Aldrich

Anti-MPI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mannose-6-phosphate isomerase, Anti-PMI, Anti-Phosphohexomutase, Anti-Phosphomannose isomerase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GSNSEVARLLASSDPLAQIAEDKPYAELWMGTHPRGDAKILDNRISQKTLSQWIAENQDSLGSKVKDTFNGNLPFLFKVLSVETPLSIQAHPNKELAEKLHLQAPQHYPDANHKPEMAIALTPFQGLCG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPI(4351)

Immunogen

Mannose-6-phosphate isomerase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MPI (Mannose phosphate isomerase) is a zinc-containing monomeric protein. It is widely distributed in human tissues including heart, brain, and skeletal muscle. It exerts phosphomannose-isomerase activity while catalyzing the interconversion reaction of fructose-6-P and mannose-6-P. It also plays a major role in providing of D-mannose derivatives at the eukaryotic glycosylation sites. Its activity is inhibited by two inhibitors, erythrose 4-phosphate and mannitol 1-phosphate. In humans, missense mutation in MPI gene causes congenital disorders of glycosylation type Ib (CDG-Ib).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71315

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Gao et al.
Analytical chemistry, 77(17), 5596-5603 (2005-09-01)
Electrospray ionization Fourier transform ion cyclotron resonance (ESI-FTICR) mass spectrometry was used to study the noncovalent complexation of a metalloenzyme, phosphomannose isomerase (PMI), which catalyzes the interconversion of mannose 6-phosphate and fructose 6-phosphate. The zinc cofactor binding effect and the
A E Proudfoot et al.
European journal of biochemistry, 219(1-2), 415-423 (1994-01-15)
Phosphomannose isomerase catalyses the interconversion of fructose-6-P and mannose-6-P and has a critical role in the supply of D-mannose derivatives required for many eukaryotic glycosylation reactions. Three classes of enzymes possessing phosphomannose-isomerase activity have been identified in bacteria and lower
E Schollen et al.
Human mutation, 16(3), 247-252 (2000-09-12)
CDG-Ib is the "gastro-intestinal" type of the congenital disorders of glycosylation (CDG) and a potentially treatable disorder. It has been described in patients presenting with congenital hepatic fibrosis and protein losing enteropathy. The symptoms result from hypoglycosylation of serum- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service