Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA017375

Sigma-Aldrich

Anti-OTUD5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-OTU domain-containing protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AAASSGLEEWTSRSPRQRSSASSPEHPELHAELGMKPPSPGTVLALAKPPSPCAPGTSSQFSAGADRATSPLVSLYPALECRALIQQMSPSAFGLNDWDDDEILASVLAVSQQEYLD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OTUD5(55593)

General description

OTU deubiquitinase 5 (OTUD5), also known as DUBA (deubiquitinating enzyme A), is a member of 14 DUBs subfamily. The protein is characterized by an ovarian tumor (OTU) domain. Phosphorylation of ser177 residue in active site will lead to the activation of enzyme.

Immunogen

OTU deubiquitinase 5

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

OTU deubiquitinase 5 (OTUD5), in association with programmed cell death 5 (PDCD5), regulates p53 mediated apoptosis through deubiquitinating mechanism. It also plays a vital role in p53 stabilization and DNA damage stress response. Increase in the expression of OTUD5 is found to efficiently enhance the activation of both PDCD5 and p53. OTUD5 bound to an adaptor protein, tumor necrosis factor receptor-associated factor 3 (TRAF3), with its ovarian tumor (OTU) domain, is involved in suppression of interferon type I (IFN-I) mediated innate immune response.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74128

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DUBA: a deubiquitinase that regulates type I interferon production.
Kayagaki N
Science, 318(5856), 1628-1632 (2007)
OTUD5 regulates p53 stability by deubiquitinating p53.
Luo J
PLoS ONE, 8(10) (2013)
Phosphorylation-dependent activity of the deubiquitinase DUBA.
Huang OW
Nature Structural and Molecular Biology, 19(2), 171-175 (2012)
Soo-Yeon Park et al.
Cancer letters, 357(1), 419-427 (2014-12-17)
Programmed cell death 5 (PDCD5) positively regulates p53-mediated apoptosis and rapidly accumulates upon DNA damage. However, the underlying mechanism of PDCD5 upregulation during the DNA damage response remains unknown. Here, we found that OTU deubiquitinase 5 (OTUD5) was bound to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service