form
liquid
shipped in
dry ice
General description
Human Hepatocyte Culture Medium is serum-free WEM-based medium for maintaining cells in culture. Base media shipped and stored refrigerated (~ 4 °C)
Application
See technical bulletins for LifeNet Health Primary Human Hepatocyte.
configured for
Product No.
Description
Pricing
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service