N0659
Noxiustoxin
≥95% (HPLC), recombinant, expressed in E. coli, lyophilized powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
recombinant
expressed in E. coli
assay
≥95% (HPLC)
form
lyophilized powder
mol wt
4195 Da
storage temp.
−20°C
Biochem/physiol Actions
Blocks voltage-gated potassium channels.
Other Notes
Originally isolated from Heteropoda venatoria
Reconstitution
Solubilize in 10 mM Tris, pH 7.5, containing 100 mM sodium chloride, 0.1% bovine serum albumin, and 1 mM EDTA
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Grissmer et al.
Molecular pharmacology, 45(6), 1227-1234 (1994-06-01)
We have analyzed the biophysical and pharmacological properties of five cloned K+ (Kv) channels (Kv1.1, Kv1.2, Kv1.3, Kv1.5, and Kv3.1) stably expressed in mammalian cell lines. Kv1.1 is biophysically similar to a K+ channel in C6 glioma cells and astrocytes
E Drakopoulou et al.
Biochemical and biophysical research communications, 213(3), 901-907 (1995-08-24)
Two forms of the Centrudoides noxius scorpion noxiustoxin, containing an amidated and an acid C-terminus, were synthesized on a solid support by using Fmoc-chemistry and 2-(1H-benzotriazol-1-yl)-1,1,3,3-tetramethyluronium hexafluorophosphate (HBTU) coupling. Comparison of the two synthetic forms with the native toxin by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service