Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAB1515

Sigma-Aldrich

Human IL-12 R beta 2 ELISA

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL12RB2(3595)

General description

The IL12RB2 (interleukin 12 receptor subunit β 2) gene is mapped to human chromosome 1p31.3 and codes for the β2 chain (130 kDa) of IL-12 receptor.
This ELISA antibody pair detects human IL-12 R beta 2. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Il12rb2 (interleukin 12 receptor subunit β 2) stimulates the release of interferon γ and Th1 (T helper cell 1) cell differentiation, as a part of autoimmune response. Il12rb2′s major function is associated with cell mediated immune system. In mice model, deletion of L12RB2 gene is associated with autoimmune and malignant disorders.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An ankylosing spondylitis-associated genetic variant in the IL23R-IL12RB2 intergenic region modulates enhancer activity and is associated with increased Th1-cell differentiation.
Roberts A R, et al.
Annals of the Rheumatic Diseases (2016)
Identification of susceptibility SNPs in IL10 and IL23R-IL12RB2 for Behcet's disease in Han Chinese.
Yu H, et al.
The Journal of Allergy and Clinical Immunology, 139(2), 621-627 (2017)
Association study of rs924080 and rs11209032 polymorphisms of IL23R-IL12RB2 in a Northern Chinese Han population with Behcet?s disease.
Qin X, et al.
Human immunology, 77(12), 1284-1290 (2016)
IL12RB2 Polymorphisms correlate with risk of lung adenocarcinoma.
Prigione I, et al.
Immunobiol., 221(2), 291-299 (2016)
Nahoko Kato-Kogoe et al.
Immunogenetics, 68(1), 55-65 (2015-11-11)
Interleukin 12 receptor β chain (IL12RB2) is a crucial regulatory factor involved in cell-mediated immune responses, and genetic variants of the gene encoding IL12RB2 are associated with susceptibility to various immune-related diseases. We previously demonstrated that haplotypes with single nucleotide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service