Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1403297

Sigma-Aldrich

Monoclonal Anti-NLRP2 antibody produced in mouse

clone 1B10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CLR19.9, FLJ20510, NALP2, NBS1, PAN1, PYPAF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~119.17 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRP2(55655)

General description

NALP proteins, such as NALP2, are characterized by an N-terminal pyrin (MIM 608107) domain (PYD) and are involved in the activation of caspase-1 (CASP1; MIM 147678) by Toll-like receptors (see TLR4; MIM 603030). They may also be involved in protein complexes that activate proinflammatory caspases (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM

Immunogen

NLRP2 (AAH39269, 1 a.a. ~ 846 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEVLTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service