Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700399

Sigma-Aldrich

Monoclonal Anti-CD18, low endotoxin antibody produced in mouse

clone MEM-148, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ITGB2, Anti-Integrin β2 subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-148, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

FUNC: suitable
flow cytometry: suitable using (QC Tested)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB2(3689)

General description

ITGB2 (integrin β2) encodes common β-chain of the β2 integrin family, known as cluster of differentiation 18 (CD18). CD18, integrin beta2 subunit, forms heterodimers with fourtypes of CD11 molecule to constitute leukocyte (beta2) integrins: alphaLbeta2(CD11a/CD18, LFA-1), alphaMbeta2 (CD11b/CD18, Mac-1, CR3), alphaXbeta2(CD11c/CD18) and alphaDbeta2 (CD11d/CD18). In most cases, the response mediatedby the integrin is a composite of the functions of its individual subunits.These integrins are essential for proper leukocyte migration, mediatingintercellular contacts. Absence of CD18 leads to leukocyte adhesiondeficiency-1; severe reduction of CD18 expression leads to the development of apsoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella)haemolytica leukotoxin and is sufficient to mediate leukotoxin-mediatedcytolysis. CD18 is primarily expressed in blood. The antibody MEM-148 recognizes an epitope on CD18 which is essentially inaccessible in intact integrin molecules on resting leukocytes, but is exposed on high-affinity state of LFA-1 or on unassociated CD18. CD18 (integrin beta2 subunit; beta2 integrin) is a 90-95 kDa type I transmembrane protein expressed on all leukocytes. In human chromosome, the gene CD18 is localized on 21q22.3.

Immunogen

Peripheral blood mononuclear cells

Application

The antibody MEM-148 induces homotypic cell aggregation and high-affinity LFA-1 conformation in resting leukocytes. Flow cytometry: Recommended dilution: 8 μg/ml. The antibody MEM-148 is an excellent marker of activated myeloid cells. Western blotting: Non-reducing conditions

Biochem/physiol Actions

Cluster of differentiation (CD18) is involved in adhesion of leukocytes to the endothelium and its migration to the site of inflammation. Lower expression of CD18 is critical because of its involvement in all the heterodimeric complex and controls the expression of α subunit. CD18 is a part of intracellular signalling molecule, signals through phosphatidylinositol 3 kinase (PI3K) and tyrosine kinase. Mutations in ITGB2 causes autosomal recessive leukocyte adhesion deficiency type 1 disorder, characterized by frequent bacterial and fungal infections. Polymorphism in ITGB2 is associated with chronic lymphocytic leukemia (CLL). Mutations in CD18 causes Hirschsprung′s disease (HSCR)-associated enterocolitis (HAEC) and chronic colitis. ITGB2 is upregulated in atherosclerotic plaque.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kindlin 3 (FERMT3) is associated with unstable atherosclerotic plaques, anti-inflammatory type II macrophages and upregulation of beta-2 integrins in all major arterial beds
Oksala N, et al.
Atherosclerosis, 242(1), 145-154 (2015)
Molecular characterization of leukocyte adhesion deficiency-I in Indian patients: identification of 9 novel mutations
Madkaikar M, et al.
Blood Cells, Molecules and Diseases, 54(3), 217-223 (2015)
Evaluating a CLL susceptibility variant in ITGB2 in families with multiple subtypes of hematological malignancies
Blackburn NB, et al.
Blood, 130(1), 86-88 (2017)
Phagocyte immunodeficiencies and their infections
Rosenzweig SD and Holland SM
The Journal of Allergy and Clinical Immunology, 113(4), 620-626 (2004)
CD11/18
D.S. Wilkes, T.J. Webb
Encyclopedia of Respiratory Medicine, 338-343 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service