Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702062

Monoclonal Anti-HLA-E purified produced in mouse

new

Synonym(s):

HLA class I histocompatibility antigen, MHC class I antigen E, alpha chain E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

MEM-E/02, monoclonal

form

buffered aqueous solution

species reactivity

( Non-human primates), human

concentration

1 mg/ml

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-E(3133)

General description

HLA-E (human leukocyte antigen E) is a non-classical MHC I antigen, which is important for dialogue with NK cells and their regulation through interaction with CD94/NKG2 receptor. Like other MHC I molecules, transmembrane HLA-E molecule (45 kDa) associates with beta2 microglobulin. Unlike HLA-G, expression of HLA-E molecules is not so restricted, but it has been detected at least at mRNA level in virtually all cells and tissues examined. In peripheral blood, HLA-E protein is expressed at least in all mononuclear cells, but in different quantity (B cells and monocytes more than T cells and NK cells).

Specificity

The antibody MEM-E/02 specifically reacts with an extracellular epitope on denaturated heavy chain of human HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on the surface of all human cell types.

Immunogen

Recombinant human HLA-E denaturated heavy chain.

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service