SAB5702375
Anti-GATA4 Antibody, clone 1M2E4, Rabbit Monoclonal
Synonym(s):
ASD2, TACHD, TOF, VSD1
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
1M2E4, monoclonal
form
liquid
mol wt
55 kDa
species reactivity
human, rat, rat
concentration
0.5 mg/mL
technique(s)
immunohistochemistry: 1:100 - 1:500
western blot: 1:500 - 1:1000
color
colorless
isotype
IgG
immunogen sequence
ASGASSNSSNATTSSSEEMRPIKTEPGLSSHYGHSSSVSQTFSVSAMSGHGPSIHPVLSALKLSPQGYASPVSQSPQTSSKQDSWNSLVLADSHGDIITA
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GATA4(2626)
General description
Immunogen
Application
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service