Skip to Content
MilliporeSigma
All Photos(8)

Documents

SAB5702824

Sigma-Aldrich

Anti-USP39 Antibody, clone 1S9R9, Rabbit Monoclonal

Synonym(s):

USP39, 65K, CGI-21, HSPC332, SAD1, SNRNP65, ubiquitin specific peptidase 39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

1S9R9, monoclonal

form

liquid

mol wt

65 kDa

species reactivity

rat, human, rat

concentration

0.3 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KNPTIVNFPITNVDLREYLSEEVQAVHKNTTYDLIANIVHDGKPSEGSYRIHVLHHGTGKWYELQDLQVTDILPQMITLSEAYIQIWKRRDNDETNQQGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP39(10713)

General description

Plays a role in pre-mRNA splicing as a component of the U4/U6-U5 tri-snRNP, one of the building blocks of the precatalytic spliceosome. Regulates AURKB mRNA levels, and thereby plays a role in cytokinesis and in the spindle checkpoint. Does not have ubiquitin-specific peptidase activity.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 466-565 of human USP39 (Q53GS9).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service