Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3525

Sigma-Aldrich

V-9302

≥98% (HPLC)

Synonym(s):

(2S)-2-Amino-4-[bis[[2-[(3-methylphenyl)methoxy]phenyl]methyl]amino]-butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H38N2O4
CAS Number:
Molecular Weight:
538.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=CC=CC(COC2=C(C=CC=C2)CN(CC3=CC=CC=C3OCC4=CC=CC(C)=C4)CC[C@@H](C(O)=O)N)=C1

InChI

1S/C34H38N2O4/c1-25-9-7-11-27(19-25)23-39-32-15-5-3-13-29(32)21-36(18-17-31(35)34(37)38)22-30-14-4-6-16-33(30)40-24-28-12-8-10-26(2)20-28/h3-16,19-20,31H,17-18,21-24,35H2,1-2H3,(H,37,38)/t31-/m0/s1

InChI key

YGKNVAAMULVFNN-HKBQPEDESA-N

Biochem/physiol Actions

V-9302 is a selective competitive antagonist of the amino acid transporter ASCT2 (SLC1A5) with anti-tumor activity. V-9302 caused reduced cellular viability and increased cell death in a panel of cancer cell lines and reduced tumor cell growth in both HCT-116 and HT29 (Fig. 5f) xenograft models.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service