SRP0162
JMJD3/KDM6B Active human
recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
Synonym(s):
JmjC domain-containing protein 3, KDM6B, KIAA0346, lysine (K)-specific demethylase 6B
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in baculovirus infected insect cells
assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
71 kDa
packaging
pkg of 20 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... KDM6B(23135)
General description
Human JMJD3/KDM6B histone demethylase (amino acids 1043-end), Genbank Accession No. NM_001080424, with C-terminal FLAG-tag, MW = 71 kDa, expressed in Sf9 cells via a baculovirus expression system.
KDM6B (lysine demethylase 6B)/ JMJD3 (jumonji domain-containing 3) is a major histone demethylase protein. This protein contains a JmjC domain in its C-terminal.
Application
Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.
Biochem/physiol Actions
KDM6B (lysine demethylase 6B) is responsible for the demethylation of histone H3K27. At promoters it demethylates the repressive H3K27me3 and therefore, promotes gene expression. In human and mouse fibroblasts, it demethyates the H3K27me3 at INK4a/ARF (alternate reading frame) locus, thus, promoting the expression of p16 protein, and therefore, acts as a tumor suppressor. It is also involved in inflammatory pathways in macrophages and embryonic stem cell differentiation. The expression of KDM6B is elevated in clear cell renal cell carcinoma (ccRCC) where it promotes EMT (epithelial to mesenchymal transition). Increased expression is also linked positively with lymph node metastasis, tumor size, cancer stage and poor prognosis.
Physical form
Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 10% glycerol and 3 mM DTT.
Preparation Note
Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
JMJD3 promotes SAHF formation in senescent WI38 cells by triggering an interplay between demethylation and phosphorylation of RB protein.
Zhao L et al
Cell Death and Differentiation, 22(10), 1630-1640 (2015)
Qiang Li et al.
International journal of clinical and experimental pathology, 8(6), 6334-6344 (2015-08-12)
Clear cell renal cell carcinoma (ccRCC) is one of the most common kidney cancers; epithelial-mesenchymal transition (EMT) is associated with carcinoma invasion and metastasis. There have been several studies about the molecular regulation of EMT, but the relationship between histone
Kristine Williams et al.
PloS one, 9(5), e96545-e96545 (2014-05-07)
The JmjC domain-containing protein JMJD3/KDM6B catalyses the demethylation of H3K27me3 and H3K27me2. JMJD3 appears to be highly regulated at the transcriptional level and is upregulated in response to diverse stimuli such as differentiation inducers and stress signals. Accordingly, JMJD3 has
JMJD3 is a histone H3K27 demethylase.
Xiang Y et al
Cell Research, 17(10), 850-857 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service