Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0260

Sigma-Aldrich

PDE2A Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

CGS-PDE, PED2A4, Phosphodiesterase 2A, cGMP-stimulated phosphodiesterase 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

assay

≥70% (SDS-PAGE)

form

aqueous solution

specific activity

≥2080 pmol/min-μg

mol wt

132 kDa

packaging

pkg of 5 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE2A(5138)

General description

Phosphodiesterase 2A (PDE2A) is encoded by the gene mapped to human chromosome 11q13.4. The encoded protein belongs to the PDE superfamily.[34}

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Biochem/physiol Actions

Phosphodiesterase 2A (PDE2A) plays a vital role in signal transduction. Activated myocardial PDE2 is considered as a potential intracellular antiadrenergic therapeutic method in heart failure (HF). The encoded protein facilitates growth and invasion of the human malignant melanoma PMP cell line. Thus, suppressing PDE2A activity might be considered as a potential method for hindering growth and invasion of malignant tumor cell lines.

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3′, 5′-cAMP to 5′ AMP per min at 30°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphodiesterase-2 is up-regulated in human failing hearts and blunts ?-adrenergic responses in cardiomyocytes.
Mehel H
Journal of the American College of Cardiology, 62, 1596-1606 (2013)
Role of phosphodiesterase 2 in growth and invasion of human malignant melanoma cells.
Hiramoto K
Cellular Signalling, 26, 1807-1817 (2014)
Integrated analysis of expression and genome alteration reveals putative amplified target genes in esophageal cancer.
Sugimoto T
Oncology Reports, 18, 465-472 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service