Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0564

Sigma-Aldrich

CD70, His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

CD27 Ligand, CD27-L, CD70 antigen, Ki-24 antigen, TNFSF7, Tumor Necrosis Factor Ligand Superfamily Member 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

19.7 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD70(970)

General description

Human CD70, also known as CD70 antigen, Tumor Necrosis Factor Ligand Superfamily Member 7, TNFSF7, CD27 Ligand, Ki-24 antigen, CD27-L, and CD27LG, GenBank Accession No. NM_001252, a.a. 39-193 with N-terminal His-tag, expressed in a HEK293 cell expression system. MW = 19.7 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service