Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3052

Sigma-Aldrich

HB-EGF human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

DTR, Diphtheria toxin receptor, HBEGF, Heparin Binding EGF-like growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in E. coli

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

1.0 ng/mL

mol wt

9.7 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HBEGF(1839)

General description

HB-EGF is an EGF related growth factor that signals through the EGF receptor, and stimulates the proliferation of smooth muscle cells (SMC), fibroblasts, epithelial cells, and keratinocytes. HB-EGF is expressed in numerous cell types and tissues, including vascular endothelial cells and SMC, macrophages, skeletal muscle, keratinocytes, and certain tumor cells. The ability of HB-EGF to specifically bind heparin and heparin sulfate proteoglycans is distinct from other EGF-like molecules, and may be related to the enhanced mitogenic activity, relative to EGF, that HB-EGF exerts on smooth muscle cells. The human HB-EGF gene encodes a 208 amino acid transmembrane protein, which can be proteolytically cleaved to produce soluble HB-EGF. Recombinant human HB-EGF is a 9.7 kDa protein containing 86 amino acid residues, corresponding to the extra-cellular EGF-like and heparin binding domains of the full length HB-EGF protein.

Application

Heparin binding EGF like growth factor has been used as a supplement in the preparation of astrocyte-enriched cultures.

Biochem/physiol Actions

HB-EGF is an EGF related growth factor that signals through the EGF receptor, and stimulates the proliferation of smooth muscle cells (SMC), fibroblasts, epithelial cells, and keratinocytes. It is involved in the autocrine or paracrine stimulation of growth of several cells and in stromal proliferation. It has been found to induce tumor growth and angiogenesis and may be a potential therapeutic target. It is also found to be essential in the development of heart and lung.

Sequence

DLQEADLDLL RVTLSSKPQA LATPNKEEHG KRKKKGKGLG KKRDPCLRKY KDFCIHGECK YVKELRAPS CICHPGYHGE RCHGLSL

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential expression of sirtuin family members in the developing, adult, and aged rat brain.
Sidorova-Darmos E
Frontiers in Aging Neuroscience, 6 (2014)
R Iwamoto et al.
Cytokine & growth factor reviews, 11(4), 335-344 (2000-08-26)
Heparin-binding EGF-like growth factor (HB-EGF), which belongs to the EGF-family growth factors, is synthesized as a membrane-anchored form (proHB-EGF). Proteolytic cleavage of proHB-EGF at the extracellular domain yields the soluble form of HB-EGF (sHB-EGF). ProHB-EGF is not only the precursor
HB-EGF is a potent inducer of tumor growth and angiogenesis.
Ongusaha PP
Cancer Research, 64, 5283-5290 (2004)
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) is a mediator of multiple physiological and pathological pathways.
Nishi E and Klagsbrun M
Growth Factors, 22, 253-260 (2004)
Fan Yang et al.
PloS one, 9(4), e94443-e94443 (2014-04-16)
Glioblastoma (GBM) is the most common primary brain tumor, accounting for approximately 40% of all central nervous system malignancies. Despite standard treatment consisting of surgical resection, radiotherapy and/or chemotherapy, the prognosis for GBM is poor; with a median survival of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service