Skip to Content
Merck
All Photos(1)

Key Documents

T8316

Sigma-Aldrich

Tertiapin

>95%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C106H176N34O23S5
CAS Number:
Molecular Weight:
2455.07
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

assay

>95%

form

powder

technique(s)

HPLC: suitable

storage temp.

−20°C

Amino Acid Sequence

Ala-Leu-Cys-Asn-Cys-Asn-Arg-Ile-Ile-Ile-Pro-His-Met-Cys-Trp-Lys-Lys-Cys-Gly-Lys-Lys-NH2

Application

Tertiapin was used for developing a novel CZE-DAD method for analyzing the honeybee venom.

Biochem/physiol Actions

A potent inhibitor of the inward-rectifier K+ channels, blocked a G-protein-gated channel (GIRK1/4) and the ROMK1 channel with nanomolar affinities, however a closely related channel, IRK1, was insensitive to tertiapin. Thus, tertiapin is a tool for purifying functional channels as well as for screening pharmaceutical agents against these channels.
Potent inhibitor of the inward-rectifier K+ channels.
Tertiapin is a peptide isolated from honey bee venom, which is a selective IKACh blocker that prolongs the atrial effective refractory period and terminates the atrial fibrillation. It is a selective and potent inhibitor of muscarinic K+ channels in rabbit cardiac myocytes.

Preparation Note

Tertiapin dissolves in water at 1 mg/ml to yield a clear solution.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zenon J Kokot et al.
Analytical and bioanalytical chemistry, 399(7), 2487-2494 (2011-01-12)
The aim of this study was to develop a new precise and accurate CZE-DAD method for honeybee venom analysis using cytochrome c as an internal standard. The 64.5 cm total length, 56 cm effective length, 75 μm ID, and 360
H Kitamura et al.
The Journal of pharmacology and experimental therapeutics, 293(1), 196-205 (2000-03-29)
Tertiapin is a 21-residue peptide isolated from honey bee venoms. A recent study indicated that tertiapin is a potent blocker of certain types of inwardly rectifying K(+) (Kir) channels (). We examined the effect of tertiapin on ion channel currents
Norio Hashimoto et al.
Pharmacological research, 54(2), 136-141 (2006-05-27)
It is well known that vagal nerve tone plays a crucial role in atrial fibrillation (AF). Acetylcholine-activated potassium current (IKACh) mediates much of the cardiac response to vagal nerve stimulation (VNS), but the contribution of IKACh to AF remains unknown.
W Jin et al.
Biochemistry, 37(38), 13291-13299 (1998-09-28)
Inward-rectifier K+ channels are a group of highly specialized K+ channels that accomplish a variety of important biological tasks. Inward-rectifier K+ channels differ from voltage-activated K+ channels not only functionally but also structurally. Each of the four subunits of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service