Skip to Content
Merck
All Photos(1)

Key Documents

Z744201

KDS Stainless Steel Syringe

new

capacity 20 mL, joint: Swagelok® 1/8, pack of 1 ea

Synonym(s):

liquid injection device, liquid withdrawal device, pump syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600

material

stainless steel

joint

joint: Swagelok® 1/8

packaging

pack of 1 ea

manufacturer/tradename

KD Scientific 780808

capacity

20 mL

Looking for similar products? Visit Product Comparison Guide

General description

Rugged stainless steel syringes are an ideal solution when the pressures and the force are high, completely eliminating the problem of breaking glass syringes. Both syringe barrel and plungers are #316 stainless steel. An O-ring seal between the top and end of the barrel protects against leakage. Syringes are guaranteed to be leak free for pressures up to 7000 psi

Application

  • High pressure applications
  • Dispensing fluids
  • Withdrawing fluids

Features and Benefits

  • Compatable with most syringe pumps
  • Eliminate hazards of glass syringe breakage
  • Reuseable - fully autoclavable
  • Adaptable to Luer Lock or Swagelok® fittings
  • Resistance to most chemicals
  • Durable construction - Electron beam welded

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service