Skip to Content
MilliporeSigma
All Photos(14)

Key Documents

ZRB1960

Sigma-Aldrich

Anti-TSC1 Antibody, clone 1C10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Hamartin;Tuberous sclerosis 1 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C10, recombinant monoclonal

description

1C10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 129.77 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TSC1(7248)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C10 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects TSC1. It targets an epitope within 20 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human TSC1.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human skeletal muscle tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected TSC1 in human skeletal muscle tissue sections.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected TSC1 in human cerebellum tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TSC1 in A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound TSC1 with a KD of 4.5 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Hamartin (UniProt: Q92574; also known as Tuberous sclerosis 1 protein, TSC1) is encoded by the TSC1 (also known as KIAA0243, TSC) gene (Gene ID: 7248) in human. TSC1 is a peripheral membrane protein that is highly expressed in skeletal muscle, followed by heart, brain, placenta, pancreas, lung, liver, and kidney. Its expression is also observed in embryonic kidney cells. In complex with TSC2, it inhibits the nutrient-mediated or growth factor-stimulated phosphorylation of S6K1 and eukaryotic translation initiation factor 4E-binding protein 1 (EIF4EBP1) by negatively regulating mTORC1 signaling. It acts as a co-chaperone for HSP90AA1 facilitating HSP90AA1 chaperoning of proteins, such as kinases, TSC2 and glucocorticoid receptor NR3C1. It increases ATP binding to HSP90AA1 and inhibits HSP90AA1 ATPase activity. It is reported to recruit TSC2 to HSP90AA1 and stabilize TSC2 by preventing the interaction between TSC2 and HECT and RLD domain containing E3 ubiquitin ligase (HERC1). It contains a C-terminal putative coiled-coil domain (721-997), which is shown to be essential for its interaction with TSC2. TSC1 and TSC2 form a protein complex (TSC1/2) that negatively regulates the mTOR pathway by inactivating RHEB. Both TSC1 and TSC2 are reported to be necessary for a functional TSC1/2 complex and loss of either gene can leads to the clinical manifestations of TSC and loss of either TSC1 or TSC2 leads to unchecked activation of mTOR. Hence, TSC1 can serve as a tumor suppressor. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Dalal, JS., et al. (2021). ELife.10; e67399; Woodford, MR., et al. (2017). EMBO. J. 36(24); 3650-3665; Tee, AR., et al. (2002). Proc. Natl. Acad. Sci. USA. 99(21); 13571-13576).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service