Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ZRB2511

Sigma-Aldrich

Anti-AMH/MIS Antibody, clone 2G11 ZooMAb® Rabbit Monoclonal

newenhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Muellerian hormone (AMH), MIF, Muellerian-inhibiting substance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 59.19 kDa
observed mol wt ~59 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey, sheep, feline, bovine, porcine, horse, canine

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... AMH(268)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2G11 is a ZooMAb® rabbit recombinant monoclonal antibody that detects AMH/MIS. It targets an epitope within 15 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human AMH/MIS.

Application

Quality Control Testing

Evaluated by Western Blotting in mouse testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AMH/MIS in mouse testis tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected AMH/MIS in mouse ovary tissue lysate.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected AMH/MIS in human ovary tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected AMH/MIS in human ovary tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with AMH/MIS peptide at least ten thousand-fold (10,000X) higher than with the non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Muellerian-inhibiting factor (UniProt: P03971; also known as Anti-Muellerian hormone (AMH), Muellerian-inhibiting substance, MIS) is encoded by the AMH (also known as MIF) gene (Gene ID: 268) in human. AMH/MIS is a glycoprotein belonging to the transforming growth factor-beta (TGF-β) superfamily. It is composed of two identical subunits linked by disulfide bonds. AMH plays a critical role in sexual differentiation by promoting the regression of the Müllerian ducts during male embryonic development, preventing the development of female reproductive structures such as the uterus and fallopian tubes. In females, AMH is involved in the regulation of folliculogenesis and ovarian function. It inhibits the initial recruitment of primordial follicles and decreases the sensitivity of growing follicles to follicle-stimulating hormone (FSH). AMH is primarily expressed in the Sertoli cells of the testes in males and the granulosa cells of ovarian follicles in females. In males, AMH expression begins during fetal development and continues until puberty, after which it decreases. In females, AMH is expressed from birth and throughout reproductive life, with levels peaking during early adulthood and declining with age. Abnormal AMH levels and signaling are associated with various reproductive disorders. In males, mutations in the AMH gene or its receptor can lead to persistent Müllerian duct syndrome (PMDS), characterized by the presence of female reproductive structures in otherwise normal males. In females, altered AMH levels are linked to conditions such as polycystic ovary syndrome (PCOS), premature ovarian insufficiency (POI), and infertility. AMH is also used as a biomarker for ovarian reserve and is measured in clinical settings to assess reproductive health and predict the onset of menopause. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service