Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ZRB2981

Sigma-Aldrich

Anti-IL-12A Antibody, clone 1G10 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Cytotoxic lymphocyte maturation factor 35 kDa subunit (CLMF p35), IL-12 subunit p35, Interleukin-12 subunit alpha, NK cell stimulatory factor chain 1 (NKSF1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 24.87 kDa
observed mol wt ~39 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

bovine, goat, monkey, porcine, sheep, baboon, canine, feline

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... IL12A(3592)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G10 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects IL-12A. It targets an epitope within 18 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal half of human IL-12A.

Application

Quality Control Testing

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected IL-12A in THP-1 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000-10,000 dilution from a representative lot detected IL-12A in lysate from human liver tissue and Raw 264.7 cells .

Flow Cytometry Analysis: 0.1 µg from a representative lot detected IL-12A in one million THP-1 cells.

Affinity Binding Assay: A representative lot of this antibody bound IL-12A peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Interleukin-12 subunit alpha (UniProt: P29459; also known as IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit (CLMF p35), IL-12 subunit p35, NK cell stimulatory factor chain 1 (NKSF1)) is encoded by the IL12A (also known as NKSF1) gene (Gene ID: 3592) in human. IL-12A is a cytokine that, along with IL12B, forms a disulfide-linked heterodimer interleukin IL-12. IL-12 alpha functions as a regulator of T cell mediated cytotoxicity, lymphocyte proliferation and activation of natural killer cells enhancing their cytotoxic activity. IL-12A is primarily localized in the cytoplasm of producing cells and is secreted into the extracellular space as part of the IL-12 heterodimer. The secretion process involves the Golgi apparatus for processing and packaging before release. IL-12A is predominantly expressed by antigen-presenting cells (APCs) such as dendritic cells, macrophages, and B cells. Its expression can be induced by various stimuli, including microbial products, antigenic stimulation, and the presence of other cytokines (e.g., IFN- ). All forms of IL-12 bind to the IL-12R which includes IL-12RbI and IL-12RbII subunits expressed on T cells and natural cells. IL-12 stimulates the production of IFN- and TNF- , subsequent production of nitric oxide that leads to pathogen destruction. Reports suggest the involvement of JAK/STAT pathway components in mediating these effects. The effects of IL-12 have been implicated in bacterial, fungal and viral infections. IL-12 is critical for controlling infections by intracellular pathogens (e.g., Mycobacterium tuberculosis). Studies have indicated that IL-35 plays an important role in controlling immune-related disorders, including autoimmune diseases, infectious diseases, and cancer. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service