All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
UNSPSC Code:
12352200
grade
primary reference standard
product line
phyproof® Reference Substance
assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
technique(s)
HPLC: suitable
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
2-8°C
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG This substance is tested for its content of d8-THC, d9-THC, and d9-THCA. Results are given on each COA. Acceptable maximum limits were set to 0.10 % each, and to 0.20 % for the sum of d9-THC and d9-THCA (in analogy to the cannabidiol monograph in DAC).
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service