1081501
USP
Butamben
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Butyl 4-aminobenzoate, Butamben
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
butamben
manufacturer/tradename
USP
bp
174 °C/8 mmHg (lit.)
mp
57-58 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CCCCOC(=O)c1ccc(N)cc1
InChI
1S/C11H15NO2/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7H,2-3,8,12H2,1H3
InChI key
IUWVALYLNVXWKX-UHFFFAOYSA-N
Gene Information
human ... TRPA1(8989) , TRPV4(59341)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monographs such as:
- Benzocaine, Butamben, and Tetracaine Hydrochloride Ointment
- Benzocaine, Butamben, and Tetracaine Hydrochloride Gel
- Benzocaine, Butamben, and Tetracaine Hydrochloride Topical Aerosol
- Benzocaine, Butamben, and Tetracaine Hydrochloride Topical Solution
Analysis Note
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service