1438318
USP
5-O-Methyvisammioside
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(S)-2-(4-Methoxy-7-methyl-5-oxo-2,3-dihydro-5H-furo[3,2-g]chromen-2-yl)propan-2-yl beta-D-glucopyranoside
Select a Size
About This Item
grade
pharmaceutical primary standard
agency
USP
packaging
pkg of 25 mg
manufacturer/tradename
USP
application(s)
pharmaceutical
format
neat
storage temp.
−20°C
1 of 4
This Item | 1000699 | 1000565 | 1134007 |
---|---|---|---|
pharmaceutical primary standard | pharmaceutical primary standard | pharmaceutical primary standard | pharmaceutical primary standard |
pharmaceutical (small molecule) | pharmaceutical (small molecule) | pharmaceutical (small molecule) | pharmaceutical (small molecule) |
neat | neat | neat | neat |
indocyanine green | acebutolol | acamprosate | - |
General description
Application
Analysis Note
Other Notes
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service