MilliporeSigma
Time since discharge of pistols and revolvers.
Journal of forensic sciences (2003-04-01)
J Andrasko, S Ståhling
ABSTRACT
The estimation of time since the latest discharge of pistols and revolvers has been achieved by the Solid Phase Microextraction (SPME) sampling technique and the GC/TEA analytical system. The TEA2 compound, which in our previous work (1) was observed in barrels of shotguns and rifles as well as inside spent cartridges, was also detected in pistols and revolvers. The amount of this compound was very low in short-barreled small arms and its escape from the barrel could generally be measured for only a few days or up to two weeks after the latest discharge. To improve the detection of TEA2 compound, the SPME sampling time was prolonged and fibers coated by Carboxen/polydimethylsiloxane were used. The decrease of the TEA2 peak with time after shooting from pistols is nonexponential but the curve-fitting procedure proposed for the estimation of time since discharge of shotguns and rifles is difficult to apply as the decay is very rapid. Therefore, the detection of TEA2 compounds in small arms should be interpreted that the firearm had been used very recently. The amount of volatile decomposition products of smokeless powder increased significantly with the length of the barrel when firearms of the same caliber, but with different barrel lengths were investigated.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μg
Estimated to ship on March 22, 2025
A recombinant, preservative-free antibody is available for your target. Try ZRB1515
$484.00