MilliporeSigma
  • Comparison of two SPME fibers for the extraction of some off-flavor cork-taint compounds in bottled wines investigated by GC-HRMS.
Comparison of two SPME fibers for the extraction of some off-flavor cork-taint compounds in bottled wines investigated by GC-HRMS.
Analytical and bioanalytical chemistry (2009-02-05)
Giuliana Bianco, Giuseppe Novario, Rosalia Zianni, Tommaso R I Cataldi
ABSTRACT
Headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography with high-resolution mass spectrometry (GC-HRMS) was used to determine 2,4,6-trichloroanisole, 2,3,6-trichloroanisole, 2,3,4-trichloroanisole, 2,3,5,6-tetrachloroanisole, pentachloroanisole, 2,4,6-tribromoanisole, 2-methylisoborneol, and 4-ethylguaiacol in wine samples. Two types of fiber coating commonly employed for sampling trichloroanisoles in wine and cork stoppers, viz. a polar mixed 50/30 microm divinylbenzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS) and a nonpolar 100 microm polydimethylsiloxane (PDMS), both 1 cm in length, were compared. This study demonstrates that the most employed polar mixed fiber should not be used with ethanol-water solutions, viz. wine samples, as its coating is not chemically and mechanically robust for sample extractions, as confirmed by environmental scanning electron microscopy. Much more effective and reliable results were obtained with the PDMS fiber, which remained functional for more than 80 analyses of red and white wine samples with satisfactory extraction efficiencies. Detection limits of investigated compounds, under optimized experimental conditions, ranged from 0.2 to 0.4 ng/L at a signal-to-noise ratio of 3 and quantification limits from 0.8 to 1.5 ng/L. The proposed method was successfully applied to commercially available Italian white and red wines using 2,4,6-TCA-d(5) as the internal standard.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
4 L
Estimated to ship on March 22, 2025
$54.40