MilliporeSigma
  • Headspace solid-phase microextraction-gas chromatographic-time-of-flight mass spectrometric methodology for geographical origin verification of coffee.
Headspace solid-phase microextraction-gas chromatographic-time-of-flight mass spectrometric methodology for geographical origin verification of coffee.
Analytica chimica acta (2008-05-20)
Sanja Risticevic, Eduardo Carasek, Janusz Pawliszyn
ABSTRACT
Increasing consumer awareness of food safety issues requires the development of highly sophisticated techniques for the authentication of food commodities. The food products targeted for falsification are either products of high commercial value or those produced in large quantities. For this reason, the present investigation is directed towards the characterization of coffee samples according to the geographical origin. The conducted research involves the development of a rapid headspace solid-phase microextraction (HS-SPME)-gas chromatography-time-of-flight mass spectrometry (GC-TOFMS) method that is utilized for the verification of geographical origin traceability of coffee samples. As opposed to the utilization of traditional univariate optimization methods, the current study employs the application of multivariate experimental designs to the optimization of extraction-influencing parameters. Hence, the two-level full factorial first-order design aided in the identification of two influential variables: extraction time and sample temperature. The optimum set of conditions for the two variables was 12 min and 55 degrees C, respectively, as directed by utilization of Doehlert matrix and response surface methodology. The high-throughput automated SPME procedure was completed by implementing a single divinylbenzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS) 50/30 microm metal fiber with excellent durability properties ensuring the completion of overall sequence of coffee samples. The utilization of high-speed TOFMS instrument ensured the completion of one GC-MS run of a complex coffee sample in 7.9 min and the complete list of benefits provided by ChromaTOF software including fully automated background subtraction, baseline correction, peak find and mass spectral deconvolution algorithms was exploited during the data evaluation procedure. The combination of the retention index (RI) system using C(8)-C(40) alkanes and the mass spectral library search was utilized for the confirmation of analyte identity in the reference authentic Brazilian coffee sample. The semi-quantitative results were then submitted to statistical evaluation, namely principal component analysis (PCA) for the establishment of geographical origin discriminations.
Product Description
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Pricing and availability is not currently available.
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Pricing and availability is not currently available.
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 1 cm
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Pricing and availability is not currently available.
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Pricing and availability is not currently available.
Supelco
ORBO-93 Sorbent Tube, ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Pricing and availability is not currently available.
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Pricing and availability is not currently available.
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Pricing and availability is not currently available.