MilliporeSigma
Vitamin D Switches BAF Complexes to Protect β Cells.
Cell (2018-05-15)
Zong Wei, Eiji Yoshihara, Nanhai He, Nasun Hah, Weiwei Fan, Antonio F M Pinto, Timothy Huddy, Yuhao Wang, Brittany Ross, Gabriela Estepa, Yang Dai, Ning Ding, Mara H Sherman, Sungsoon Fang, Xuan Zhao, Christopher Liddle, Annette R Atkins, Ruth T Yu, Michael Downes, Ronald M Evans
ABSTRACT
A primary cause of disease progression in type 2 diabetes (T2D) is β cell dysfunction due to inflammatory stress and insulin resistance. However, preventing β cell exhaustion under diabetic conditions is a major therapeutic challenge. Here, we identify the vitamin D receptor (VDR) as a key modulator of inflammation and β cell survival. Alternative recognition of an acetylated lysine in VDR by bromodomain proteins BRD7 and BRD9 directs association to PBAF and BAF chromatin remodeling complexes, respectively. Mechanistically, ligand promotes VDR association with PBAF to effect genome-wide changes in chromatin accessibility and enhancer landscape, resulting in an anti-inflammatory response. Importantly, pharmacological inhibition of BRD9 promotes PBAF-VDR association to restore β cell function and ameliorate hyperglycemia in murine T2D models. These studies reveal an unrecognized VDR-dependent transcriptional program underpinning β cell survival and identifies the VDR:PBAF/BAF association as a potential therapeutic target for T2D.
Sigma-Aldrich
Pipecolinic acid, 98%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Pricing and Inventory is not available. Please check the Millipore website for more information.
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Histopaque®-1077 Hybri-Max, liquid, sterile-filtered, BioReagent, suitable for hybridoma
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Bovine INS / Insulin ELISA Kit
Pricing and Inventory is not available. Please check the Millipore website for more information.
Millipore
ANTI-FLAG® High Sensitivity, M2 coated 96-well plates, 96-well, clear, polystyrene, flat bottom plate
Pricing and Inventory is not available. Please check the Millipore website for more information.