MilliporeSigma
  • Spin-Center Shift-Enabled Direct Enantioselective α-Benzylation of Aldehydes with Alcohols.
Spin-Center Shift-Enabled Direct Enantioselective α-Benzylation of Aldehydes with Alcohols.
Journal of the American Chemical Society (2018-02-06)
Eric D Nacsa, David W C MacMillan
ABSTRACT
Nature routinely engages alcohols as leaving groups, as DNA biosynthesis relies on the removal of water from ribonucleoside diphosphates by a radical-mediated "spin-center shift" (SCS) mechanism. Alcohols, however, remain underused as alkylating agents in synthetic chemistry due to their low reactivity in two-electron pathways. We report herein an enantioselective α-benzylation of aldehydes using alcohols as alkylating agents based on the mechanistic principle of spin-center shift. This strategy harnesses the dual activation modes of photoredox and organocatalysis, engaging the alcohol by SCS and capturing the resulting benzylic radical with a catalytically generated enamine. Mechanistic studies provide evidence for SCS as a key elementary step, identify the origins of competing reactions, and enable improvements in chemoselectivity by rational photocatalyst design.
Sigma-Aldrich
Ir(p-tBu-ppy)3