MilliporeSigma
  • Localization of αA-Crystallin in Rat Retinal Müller Glial Cells and Photoreceptors.
Localization of αA-Crystallin in Rat Retinal Müller Glial Cells and Photoreceptors.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada (2018-09-27)
Astrid Zayas-Santiago, David S Ríos, Lidia V Zueva, Mikhail Y Inyushin
ABSTRACT
Transparent cells in the vertebrate optical tract, such as lens fiber cells and corneal epithelium cells, have specialized proteins that somehow permit only a low level of light scattering in their cytoplasm. It has been shown that both cell types contain (1) beaded intermediate filaments as well as (2) α-crystallin globulins. It is known that genetic and chemical alterations to these specialized proteins induce cytoplasmic opaqueness and visual complications. Crystallins were described previously in the retinal Müller cells of frogs. In the present work, using immunocytochemistry, fluorescence confocal imaging, and immuno-electron microscopy, we found that αA-crystallins are present in the cytoplasm of retinal Müller cells and in the photoreceptors of rats. Given that Müller glial cells were recently described as "living light guides" as were photoreceptors previously, we suggest that αA-crystallins, as in other highly transparent cells, allow Müller cells and photoreceptors to minimize intraretinal scattering during retinal light transmission.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)−Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 5 nm (colloidal gold)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Triton X-100, BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00
Sigma-Aldrich
Triton X-100, laboratory grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Estimated to ship on March 22, 2025
$187.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
0.1 mL
Estimated to ship on March 22, 2025
$214.00
7 mL
Estimated to ship on March 22, 2025
$472.00
0.5 mL
Estimated to ship on March 22, 2025
$617.00
1 mL
Estimated to ship on March 22, 2025
$953.00