MilliporeSigma
  • Pharmacological evidence for the 5-HT7 receptor mediating smooth muscle relaxation in canine cerebral arteries.
Pharmacological evidence for the 5-HT7 receptor mediating smooth muscle relaxation in canine cerebral arteries.
British journal of pharmacology (1999-07-13)
J A Terrón, A Falcón-Neri
ABSTRACT
1. We investigated in the present study whether 5-HT is able to exert direct relaxant responses in canine basilar and middle cerebral arteries via the 5-HT7 receptor. 2. In arterial rings deprived of endothelium and pre-contracted with prostaglandin F2 alpha (2 microM), 5-HT, 5-carboxamidotryptamine (5-CT), 5-methoxytryptamine, sumatriptan or alpha-methyl-5-HT produced further increase in tone and/or slight relaxation. Blockade of 5-HT1B 1D and 5-HT2A receptors with GR127935 (1 microM) and ketanserin (0.1 microM), respectively, antagonized the vasoconstrictor component of the response and unmasked a concentration-dependent relaxation to 5-HT, 5-CT and 5-methoxytryptamine; sumatriptan and alpha-methyl-5-HT remained inactive as relaxant agonists. The rank order of agonist potency in both arteries was 5-CT > 5-HT > 5-methoxytryptamine > sumatriptan > or = alpha-methyl-5-HT. 3. In dog basilar artery, pre-incubated with GR127935 (1 microM) and ketanserin (0.1 microM) and precontracted with prostaglandin F2 alpha (2 microM), the 5-HT7 ligands, clozapine (1 microM), mesulergine (0.3 microM), methiothepin (3 nM), risperidone (3 nM), spiperone (1 microM) and LY215840 (10-100 nM), produced significant rightward shifts of the concentration-response curves for 5-HT and 5-CT. Only methiothepin and risperidone reduced significantly the maximum relaxant response (Emax), whilst the other drugs behaved as competitive antagonists with affinity values (pKB) that significantly correlated with their binding affinity (pKi) at recombinant 5-HT7 receptors. 4. These data disclosing the involvement of the 5-HT7 receptor in cerebrovascular relaxation may be strongly relevant in the light of: (1) the involvement of 5-HT in migraine; (2) the putative linkage between cephalovascular vasodilatation and migraine headache; and (3) the relatively high 5-HT7 receptor affinity of migraine prophylactic 5-HT antagonists.
Sigma-Aldrich
ChemiSCREEN Membrane Preparation Recombinant Human 5-HT₇ Serotonin Receptor, Human 5-HT7 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on March 23, 2025
$135.00