MilliporeSigma
  • Theory and practice of combining coagulant fixation and microwave histoprocessing.
Theory and practice of combining coagulant fixation and microwave histoprocessing.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2008-11-26)
M E Boon, L P Kok
ABSTRACT
The German, F. Blum, introduced formalin as a fixative in 1893. Formalin rapidly became popular for hardening and preserving gross human and animal specimens. As a result, microscopy for diagnostic pathology by combining paraffin embedding and formalin fixation was developed. Alcohol-based fixatives have coagulation of proteins as their main preservative effect. Because there is no cross-linking, immunostaining is not compromised, and DNA and RNA is not damaged. Ethyl alcohol was used by Dutch scientists of the 18th century, but was replaced by the cheaper formalin. Addition of low molecular weight polyethylene glycol (PEG) optimized the coagulant fixative, Kryofix. The polyethylene glycol prevents excessive hardening and enhances the speed of coagulation of proteins. Kryofix was used on a large scale for skin biopsies in Leiden between 1987 and 2001. DNA preservation by the formulated coagulant fixative, BoonFix, is related to the concentration of ethyl alcohol, PEG and acetic acid. BoonFix has been used since 2004 in Leiden for over 40,000 diagnostic skin biopsies and more than 100,000 cervical samples. A literature review and three decades of experience with coagulant, formalin-free fixatives in pathology suggest that when health authorities realize that formalin invalidates expensive tests, it might eventually be eliminated legislatively from diagnostic pathology. Finally, coagulant fixation is optimal for microwave histoprocessing where ethyl alcohol is followed by isopropanol.
Sigma-Aldrich
Formalin Free Tissue Fixative
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
₩136,112
100 g
In Stock
Details...
₩374,028