MilliporeSigma
  • Epigenetic inhibitor zebularine activates ear pinna wound closure in the mouse.
Epigenetic inhibitor zebularine activates ear pinna wound closure in the mouse.
EBioMedicine (2019-07-16)
Piotr Sass, Paweł Sosnowski, Justyna Podolak-Popinigis, Bartosz Górnikiewicz, Jolanta Kamińska, Milena Deptuła, Ewa Nowicka, Anna Wardowska, Jarosław Ruczyński, Piotr Rekowski, Piotr Rogujski, Natalia Filipowicz, Alina Mieczkowska, Grażyna Peszyńska-Sularz, Łukasz Janus, Piotr Skowron, Artur Czupryn, Piotr Mucha, Arkadiusz Piotrowski, Sylwia Rodziewicz-Motowidło, Michał Pikuła, Paweł Sachadyn
ABSTRACT
Most studies on regenerative medicine focus on cell-based therapies and transplantations. Small-molecule therapeutics, though proved effective in different medical conditions, have not been extensively investigated in regenerative research. It is known that healing potential decreases with development and developmental changes are driven by epigenetic mechanisms, which suggests epigenetic repression of regenerative capacity. We applied zebularine, a nucleoside inhibitor of DNA methyltransferases, to stimulate the regenerative response in a model of ear pinna injury in mice. We observed the regeneration of complex tissue that was manifested as improved ear hole repair in mice that received intraperitoneal injections of zebularine. Six weeks after injury, the mean hole area decreased by 83.2 ± 9.4% in zebularine-treated and by 43.6 ± 15.4% in control mice (p < 10-30). Combined delivery of zebularine and retinoic acid potentiated and accelerated this effect, resulting in complete ear hole closure within three weeks after injury. We found a decrease in DNA methylation and transcriptional activation of neurodevelopmental and pluripotency genes in the regenerating tissues. This study is the first to demonstrate an effective induction of complex tissue regeneration in adult mammals using zebularine. We showed that the synergistic action of an epigenetic drug (zebularine) and a transcriptional activator (retinoic acid) could be effectively utilized to induce the regenerative response, thus delineating a novel pharmacological strategy for regeneration. The strategy was effective in the model of ear pinna regeneration in mice, but zebularine acts on different cell types, therefore, a similar approach can be tested in other tissues and organs.
Product Description
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Zebularine, ≥98% (HPLC), solid