MilliporeSigma
  • Herpes simplex viruses activate phospholipid scramblase to redistribute phosphatidylserines and Akt to the outer leaflet of the plasma membrane and promote viral entry.
Herpes simplex viruses activate phospholipid scramblase to redistribute phosphatidylserines and Akt to the outer leaflet of the plasma membrane and promote viral entry.
PLoS pathogens (2018-01-03)
Natalia Cheshenko, Carl Pierce, Betsy C Herold
ABSTRACT
Herpes simplex virus (HSV) entry is associated with Akt translocation to the outer leaflet of the plasma membrane to promote a complex signaling cascade. We hypothesized that phospholipid scramblase-1 (PLSCR1), a calcium responsive enzyme that flips phosphatidylserines between membrane leaflets, might redistribute Akt to the outside during entry. Confocal imaging, biotinylation of membrane proteins and flow cytometric analysis demonstrated that HSV activates PLSCR1 and flips phosphatidylserines and Akt to the outside shortly following HSV-1 or HSV-2 exposure. Translocation was blocked by addition of a cell permeable calcium chelator, pharmacological scramblase antagonist, or transfection with small interfering RNA targeting PLSCR1. Co-immunoprecipitation and proximity ligation studies demonstrated that PLSCR1 associated with glycoprotein L at the outer leaflet and studies with gL deletion viruses indicate that this interaction facilitates subsequent restoration of the plasma membrane architecture. Ionomycin, a calcium ionophore, also induced PLSCR1 activation resulting in Akt externalization, suggesting a previously unrecognized biological phenomenon.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, 4G10® Platinum, clone 4G10, Upstate®, from mouse
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
Anti-Phosphatidylserine Antibody, clone 1H6, clone 1H6, Upstate®, from mouse