MilliporeSigma
  • Optimized Isolation of Extracellular Vesicles From Various Organic Sources Using Aqueous Two-Phase System.
Optimized Isolation of Extracellular Vesicles From Various Organic Sources Using Aqueous Two-Phase System.
Scientific reports (2019-12-18)
Oğuz Kaan Kırbaş, Batuhan Turhan Bozkurt, Ayla Burçin Asutay, Beyza Mat, Bihter Ozdemir, Dilek Öztürkoğlu, Hülya Ölmez, Zeynep İşlek, Fikrettin Şahin, Pakize Neslihan Taşlı
ABSTRACT
From biomarkers to drug carriers, Extracellular Vesicles (EVs) are being used successfully in numerous applications. However, while the subject has been steadily rising in popularity, current methods of isolating EVs are lagging behind, incapable of isolating EVs at a high enough quantity or quality while also requiring expensive, specialized equipment. The "isolation problem" is one of the major obstacles in the field of EV research - and even more so for their potential, widespread use for clinical diagnosis and therapeutic applications. Aqueous Two-Phase Systems (ATPS) has been reported previously as a promising method for isolating EVs quickly and efficiently, and with little contaminants - however, this method has not seen widespread use. In this study, an ATPS-based isolation protocol is used to isolate small EVs from plant, cell culture, and parasite culture sources. Isolated EVs were characterized in surface markers, size, and morphological manner. Additionally, the capacity of ATPS-based EV isolation in removing different contaminants was shown by measuring protein, fatty acid, acid, and phenol red levels of the final isolate. In conclusion, we have shown that EVs originating from different biological sources can be isolated successfully in a cost-effective and user-friendly manner with the use of aqueous two-phase systems.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Poly(ethylene glycol), 35,000