MilliporeSigma
  • Post-surgical adhesions are triggered by calcium-dependent membrane bridges between mesothelial surfaces.
Post-surgical adhesions are triggered by calcium-dependent membrane bridges between mesothelial surfaces.
Nature communications (2020-06-20)
Adrian Fischer, Tim Koopmans, Pushkar Ramesh, Simon Christ, Maximilian Strunz, Juliane Wannemacher, Michaela Aichler, Annette Feuchtinger, Axel Walch, Meshal Ansari, Fabian J Theis, Kenji Schorpp, Kamyar Hadian, Philipp-Alexander Neumann, Herbert B Schiller, Yuval Rinkevich
ABSTRACT
Surgical adhesions are bands of scar tissues that abnormally conjoin organ surfaces. Adhesions are a major cause of post-operative and dialysis-related complications, yet their patho-mechanism remains elusive, and prevention agents in clinical trials have thus far failed to achieve efficacy. Here, we uncover the adhesion initiation mechanism by coating beads with human mesothelial cells that normally line organ surfaces, and viewing them under adhesion stimuli. We document expansive membrane protrusions from mesothelia that tether beads with massive accompanying adherence forces. Membrane protrusions precede matrix deposition, and can transmit adhesion stimuli to healthy surfaces. We identify cytoskeletal effectors and calcium signaling as molecular triggers that initiate surgical adhesions. A single, localized dose targeting these early germinal events completely prevented adhesions in a preclinical mouse model, and in human assays. Our findings classifies the adhesion pathology as originating from mesothelial membrane bridges and offer a radically new therapeutic approach to treat adhesions.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Weigert′s Iron Hematoxylin Set, Set
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Trichrome Stain (Masson) Kit
Sigma-Aldrich
Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Bouin′s solution, histological fixative
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer