MilliporeSigma
  • Cancer Burden Is Controlled by Mural Cell-β3-Integrin Regulated Crosstalk with Tumor Cells.
Cancer Burden Is Controlled by Mural Cell-β3-Integrin Regulated Crosstalk with Tumor Cells.
Cell (2020-05-31)
Ping-Pui Wong, José M Muñoz-Félix, Maruan Hijazi, Hyojin Kim, Stephen D Robinson, Beatriz De Luxán-Delgado, Irene Rodríguez-Hernández, Oscar Maiques, Ya-Ming Meng, Qiong Meng, Natalia Bodrug, Matthew Scott Dukinfield, Louise E Reynolds, George Elia, Andrew Clear, Catherine Harwood, Yu Wang, James J Campbell, Rajinder Singh, Penglie Zhang, Thomas J Schall, Kylie P Matchett, Neil C Henderson, Peter W Szlosarek, Sally A Dreger, Sally Smith, J Louise Jones, John G Gribben, Pedro R Cutillas, Pascal Meier, Victoria Sanz-Moreno, Kairbaan M Hodivala-Dilke
ABSTRACT
Enhanced blood vessel (BV) formation is thought to drive tumor growth through elevated nutrient delivery. However, this observation has overlooked potential roles for mural cells in directly affecting tumor growth independent of BV function. Here we provide clinical data correlating high percentages of mural-β3-integrin-negative tumor BVs with increased tumor sizes but no effect on BV numbers. Mural-β3-integrin loss also enhances tumor growth in implanted and autochthonous mouse tumor models with no detectable effects on BV numbers or function. At a molecular level, mural-cell β3-integrin loss enhances signaling via FAK-p-HGFR-p-Akt-p-p65, driving CXCL1, CCL2, and TIMP-1 production. In particular, mural-cell-derived CCL2 stimulates tumor cell MEK1-ERK1/2-ROCK2-dependent signaling and enhances tumor cell survival and tumor growth. Overall, our data indicate that mural cells can control tumor growth via paracrine signals regulated by β3-integrin, providing a previously unrecognized mechanism of cancer growth control.
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Pimonidazole, ≥98% (HPLC)
Sigma-Aldrich
Anti-NG2, Alexa Fluor488 Conjugate Antibody, from rabbit, ALEXA FLUOR 488