MilliporeSigma
  • Domain analysis of lipoprotein LppQ in Mycoplasma mycoides subsp. mycoides SC.
Domain analysis of lipoprotein LppQ in Mycoplasma mycoides subsp. mycoides SC.
Antonie van Leeuwenhoek (2007-08-04)
Laetitia Bonvin-Klotz, Edy M Vilei, Kathrin Kühni-Boghenbor, Nadine Kapp, Joachim Frey, Michael H Stoffel
ABSTRACT
The lipoprotein LppQ is the most prominent antigen of Mycoplasma mycoides subsp. mycoides small colony type (SC) during infection of cattle. This pathogen causes contagious bovine pleuropneumonia (CBPP), a devastating disease of considerable socio-economic importance in many countries worldwide. The dominant antigenicity and high specificity for M. mycoides subsp. mycoides SC of lipoprotein LppQ have been exploited for serological diagnosis and for epidemiological investigations of CBPP. Scanning electron microscopy and immunogold labelling were used to provide ultrastructural evidence that LppQ is located to the cell membrane at the outer surface of M. mycoides subsp. mycoides SC. The selectivity and specificity of this method were demonstrated through discriminating localization of extracellular (i.e., in the zone of contact with host cells) vs. integral membrane domains of LppQ. Thus, our findings support the suggestion that the accessible N-terminal domain of LppQ is surface exposed and such surface localization may be implicated in the pathogenesis of CBPP.
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$157.00
$136.59
500 mL
In Stock
Details...
$539.00
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
In Stock
Details...
$155.00
5 kg
In Stock
Details...
$758.00
20 kg
Estimated to ship on March 24, 2025
$2,640.00
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mL
In Stock
Details...
$79.10
250 mL
Estimated to ship on March 24, 2025
$327.00
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mL
In Stock
Details...
$125.00
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
In Stock
Details...
$85.20
1 L
In Stock
Details...
$365.00
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mL
In Stock
Details...
$51.60
100 mL
In Stock
Details...
$114.00