Merck
  • Comparative carcinogenicity study of a thick, straight-type and a thin, tangled-type multi-walled carbon nanotube administered by intra-tracheal instillation in the rat.
Comparative carcinogenicity study of a thick, straight-type and a thin, tangled-type multi-walled carbon nanotube administered by intra-tracheal instillation in the rat.
Particle and fibre toxicology (2020-10-16)
Dina Mourad Saleh, William T Alexander, Takamasa Numano, Omnia Hosny Mohamed Ahmed, Sivagami Gunasekaran, David B Alexander, Mohamed Abdelgied, Ahmed M El-Gazzar, Hiroshi Takase, Jiegou Xu, Aya Naiki-Ito, Satoru Takahashi, Akihiko Hirose, Makoto Ohnishi, Jun Kanno, Hiroyuki Tsuda
ABSTRACT
Multi-walled carbon nanotubes can be divided into two general subtypes: tangled and straight. MWCNT-N (60 nm in diameter) and MWCNT-7 (80-90 nm in diameter) are straight-type MWCNTs, and similarly to asbestos, both are carcinogenic to the lung and pleura when administered to rats via the airway. Injection of straight-type MWCNTs into the peritoneal cavity also induces the development of mesothelioma, however, injection of tangled-type MWCNTs into the peritoneal cavity does not induce carcinogenesis. To investigate these effects in the lung we conducted a 2-year comparative study of the potential carcinogenicities of a straight-type MWCNT, MWCNT-A (approximately 150 nm in diameter), and a tangled-type MWCNT, MWCNT-B (7.4 nm in diameter) after administration into the rat lung. Crocidolite asbestos was used as the reference material, and rats administered vehicle were used as the controls. Test materials were administered by intra-Tracheal Intra-Pulmonary Spraying (TIPS) once a week over a 7 week period (8 administrations from day 1 to day 50), followed by a 2-year observation period without further treatment. Rats were administered total doses of 0.5 or 1.0 mg MWCNT-A and MWCNT-B or 1.0 mg asbestos. There was no difference in survival between any of the groups. The rats administered MWCNT-A or asbestos did not have a significant increase in bronchiolo-alveolar hyperplasia or tumors in the lung. However, the rats administered MWCNT-B did have significantly elevated incidences of bronchiolo-alveolar hyperplasia and tumors in the lung: the incidence of bronchiolo-alveolar hyperplasia was 0/20, 6/20, and 9/20 in the vehicle, 0.5 mg MWCNT-B, and 1.0 mg MWCNT-B groups, respectively, and the incidence of adenoma and adenocarcinoma combined was 1/19, 5/20, and 7/20 in the vehicle, 0.5 mg MWCNT-B, and 1.0 mg MWCNT-B groups, respectively. Malignant pleural mesothelioma was not induced in any of the groups. The results of this initial study indicate that tangled-type MWCNT-B is carcinogenic to the rat lung when administered via the airway, and that straight-type MWCNT-A did not have higher carcinogenic potential in the rat lung than tangled-type MWCNT-B.
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Pricing and availability is not currently available.