Merck
  • Growth inhibition of pathogenic microorganisms by Pseudomonas protegens EMM-1 and partial characterization of inhibitory substances.
Growth inhibition of pathogenic microorganisms by Pseudomonas protegens EMM-1 and partial characterization of inhibitory substances.
PloS one (2020-10-16)
Catherine Cesa-Luna, Antonino Baez, Alberto Aguayo-Acosta, Roberto Carlos Llano-Villarreal, Víctor Rivelino Juárez-González, Paul Gaytán, María Del Rocío Bustillos-Cristales, América Rivera-Urbalejo, Jesús Muñoz-Rojas, Verónica Quintero-Hernández
ABSTRACT
The bacterial strain, EMM-1, was isolated from the rhizosphere of red maize ("Rojo Criollo") and identified as Pseudomonas protegens EMM-1 based on phylogenetic analysis of 16S rDNA, rpoB, rpoD, and gyrB gene sequences. We uncovered genes involved in the production of antimicrobial compounds like 2,4-diacetylphloroglucinol (2,4-DAPG), pyoluteorin, and lectin-like bacteriocins. These antimicrobial compounds are also produced by other fluorescent pseudomonads alike P. protegens. Double-layer agar assay showed that P. protegens EMM-1 inhibited the growth of several multidrug-resistant (MDR) bacteria, especially clinical isolates of the genera Klebsiella and β-hemolytic Streptococcus. This strain also displayed inhibitory effects against diverse fungi, such as Aspergillus, Botrytis, and Fusarium. Besides, a crude extract of inhibitory substances secreted into agar was obtained after the cold-leaching process, and physicochemical characterization was performed. The partially purified inhibitory substances produced by P. protegens EMM-1 inhibited the growth of Streptococcus sp. and Microbacterium sp., but no inhibitory effect was noted for other bacterial or fungal strains. The molecular weight determined after ultrafiltration was between 3 and 10 kDa. The inhibitory activity was thermally stable up to 60°C (but completely lost at 100°C), and the inhibitory activity remained active in a wide pH range (from 3 to 9). After treatment with a protease from Bacillus licheniformis, the inhibitory activity was decreased by 90%, suggesting the presence of proteic natural compounds. All these findings suggested that P. protegens EMM-1 is a potential source of antimicrobials to be used against pathogens for humans and plants.
Sigma-Aldrich
Protease from Bacillus licheniformis, ≥2.4 U/g