MilliporeSigma
Mediation of the Acute Stress Response by the Skeleton.
Cell metabolism (2019-09-17)
Julian Meyer Berger, Parminder Singh, Lori Khrimian, Donald A Morgan, Subrata Chowdhury, Emilio Arteaga-Solis, Tamas L Horvath, Ana I Domingos, Anna L Marsland, Vijay Kumar Yadav, Kamal Rahmouni, Xiao-Bing Gao, Gerard Karsenty
ABSTRACT
We hypothesized that bone evolved, in part, to enhance the ability of bony vertebrates to escape danger in the wild. In support of this notion, we show here that a bone-derived signal is necessary to develop an acute stress response (ASR). Indeed, exposure to various types of stressors in mice, rats (rodents), and humans leads to a rapid and selective surge of circulating bioactive osteocalcin because stressors favor the uptake by osteoblasts of glutamate, which prevents inactivation of osteocalcin prior to its secretion. Osteocalcin permits manifestations of the ASR to unfold by signaling in post-synaptic parasympathetic neurons to inhibit their activity, thereby leaving the sympathetic tone unopposed. Like wild-type animals, adrenalectomized rodents and adrenal-insufficient patients can develop an ASR, and genetic studies suggest that this is due to their high circulating osteocalcin levels. We propose that osteocalcin defines a bony-vertebrate-specific endocrine mediation of the ASR.
Sigma-Aldrich
Gastrin EIA Kit, for serum, plasma, culture supernatant and cell lysates
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody, from goat, purified by affinity chromatography
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Protein Gene Product 9.5 Antibody, serum, Chemicon®