Merck
  • Codon Deoptimization of UL54 in meq-Deleted Marek's Disease Vaccine Candidate Eliminates Lymphoid Atrophy but Reduces Vaccinal Protection.
Codon Deoptimization of UL54 in meq-Deleted Marek's Disease Vaccine Candidate Eliminates Lymphoid Atrophy but Reduces Vaccinal Protection.
Avian diseases (2020-11-19)
Steven J Conrad, Cari J Hearn, Robert F Silva, John R Dunn
ABSTRACT
Marek's disease (MD) is an oncogenic, lymphoproliferative, and highly contagious disease of chickens. Its etiologic agent is the alphaherpesvirus Marek's disease virus (MDV, Gallid alphaherpesvirus 2), and it is a chronic and ubiquitous problem for the poultry industry with significant economic impact in the United States and worldwide. We have previously demonstrated that MDV attenuated by dicodon deoptimization of the UL54 gene results in reduced gene product accumulation in vitro, with reduced viral genome copy number upon infection and reduced atrophy of bursa and thymus in vivo as well. In this report we detail our attempts to use the same attenuation strategy on a meq-deleted MDV mutant, rMd5B40ΔMeq. Unlike the wild-type rMd5B40 virus the rMd5B40ΔMeq is no longer oncogenic, but infected birds experience an unacceptable amount of bursa and thymus atrophy (BTA). We produced two meq-deleted MDV recombinants with a dicodon-deoptimized UL54 (rMd5B40ΔMeq/UL54deop1 and -deop2) and tested their tendency to cause BTA and to serve as a protective vaccine. We found that, although dicodon deoptimization of the UL54 gene results in a virus that spares the infected animal from atrophy of the bursa and thymus, the meq-deleted UL54-deoptimized recombinant is also less protective than the meq-deleted virus without UL54 deoptimization, the HVT + SB1 combination vaccine, or the Rispens (CVI988) vaccine.
Sigma-Aldrich
L-15 Medium (Leibovitz), With L-glutamine, powder, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
McCoy′s 5A Medium, Modified, with L-glutamine, without sodium bicarbonate, powder, suitable for cell culture